Learn More
Selleck Chemical LLC Apoptozole 10mg 1054543-47-3 Apoptosis Activator VII
SDP
Catalog No. 501365986
Encompass_Preferred
Apoptozole (Apoptosis Activator VII) is an inhibitor of heat shock protein 70(HSP70) and Hsc70 with dissociation constants Kd of 0.14 ?M and 0.21 ?M, respectively. It induces caspase-dependent apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5986 Supplier Selleck Chemical LLC Supplier No. S836510MG
Apoptozole (Apoptosis Activator VII) is an inhibitor of heat shock protein 70(HSP70) and Hsc70 with dissociation constants Kd of 0.14 ?M and 0.21 ?M, respectively. It induces caspase-dependent apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.