Learn More
Cayman Chemical AposcopolamIn 10mg
SDP
Catalog No. 502909514
Encompass_Preferred
A tropane alkaloid and an active metabolite of scopolamine; selective for mAChRs over nAChRs (IC50s = 0.0192 and 188 µM, respectively)
Catalog No. 50-290-9514 Supplier Cayman Chemical Supplier No. 3668910 MG
A tropane alkaloid and an active metabolite of scopolamine; selective for mAChRs over nAChRs (IC50s = 0.0192 and 188 µM, respectively)