Learn More
Aviva Systems Biology Corp APP Antibody - middle region (ARP33074_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33074_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human APP. Alternate names: AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN2, PN-IIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54874