Learn More
Aviva Systems Biology Corp App Antibody - C-terminal region (ARP58851_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58851_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against App. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human App. Alternate names: Abeta, Abpp, Adap, Ag, betaApp, Cvap, E030013M08RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-503