Learn More
IBL America APP βCTF
Supplier:  IBL America 27776
Encompass_Preferred
Solid phase sandwich ELISA for the determination of human APP CFT in cell lysate. For research use only, not for use in diagnostic procedures.This ELISA can measure human APP CTF in samples such as cultured cell lysate.Tetra Methyl Benzidine (TMB) is used as coloring agent (Chromogen). The strength of coloring is in proportion to the quantities of human APP CTF.
Catalog No. 50-136-6559