Learn More
Enzo Life Sciences APP monoclonal antibody (1G6) (200µl)
SDP
Supplier:  Enzo Life Sciences ENZABS1280200
Encompass_Preferred
Clone: 1G6. Isotype: IgG2b. Species reactivity: Human. Specificity: Epitope lies within amino acids 573-596 of amyloid precursor protein. Applications: ELISA, IHC (PS), WB. Purity: Purified. Formulation: Liquid. In PBS. Does not contain any preservatives or carrier proteins.
Catalog No. 50-201-2692