Learn More
Enzo Life Sciences APP monoclonal antibody (3E9) (100µg)
SDP
Supplier:  Enzo Life Sciences ADINBA100E
Encompass_Preferred
Clone: 3E9. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of human APP695. UniProt ID: P05067. Species reactivity: Human, Mouse. Applications: WB. Recommended Dilutions: Western Blot (5-10µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid.  In PBS pH7.2 containing 50% glycerol.
Catalog No. 50-200-7353