Learn More
Enzo Life Sciences APP monoclonal antibody (D3B10) (200µl)
SDP
Supplier:  Enzo Life Sciences ENZABS1450200
Encompass_Preferred
Clone: D3B10. Isotype: IgG1. Species reactivity: Human. Specificity: Epitope lies within amino acids 61-77 of amyloid precursor protein. Applications: ELISA, IHC (PS), WB. Purity: Protein A purified. Formulation: Liquid. In PBS. Does not contain any preservatives or carrier proteins.
Catalog No. 50-201-2705