Learn More
Enzo Life Sciences APP monoclonal antibody (LN27) (200µl)
SDP
Supplier:  Enzo Life Sciences ENZABS1260200
Encompass_Preferred
Clone: LN27. Isotype: IgG1. Species reactivity: Human. Specificity: Recognizes all three known forms of APP (β-APP695, β-APP751, β-APP770). Epitope lies within amino acids 45-53 of amyloid precursor protein. Applications: WB. Purity: Protein G purified. Formulation: Liquid. In PBS. Does not contain any preservatives or carrier proteins.
Catalog No. 50-201-2688