Learn More
Enzo Life Sciences APP polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADINBA102E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 737-751 of human APP or aa 85-99 of human C99 (a fragment generated by β-secretase cleavage). The sequence of this peptide is completely conserved in mouse and rat APP. Species reactivity: Human, Mouse, Rat. Applications: IHC, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-7354