Learn More
Enzo Life Sciences APP polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2260100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human APP. Source: Purified from rabbit serum. Species reactivity: Human, Mouse. Applications: ELISA, IP, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-201-2775