Learn More
Waters Corp Waters ACQUITY Premier Protein SEC Column, 250 Å, 1.7 μm, 4.6 x 300 mm
Supplier:  Waters Corp 186009964
Encompass_Preferred
ACQUITY Premier Protein SEC Columns deliver reliable protein size variant separations (10,000 to 650,000 Daltons). Combining MaxPeak Premier High Performance Surfaces with BEH-PEO Particle technologies, these columns minimize undesirable secondary ionic or hydrophobic protein interactions.
  • Minimizes undesirable secondary ionic/hydrophobic protein interactions
  • Reduces method development time with generic methodologies
  • Batch QC tested for consistent protein separation performance
  • Optimized for UPLC SEC applications with low LC system volume
  • Supports gel filtration and size exclusion chromatography
  • Analyzes aggregates, monomers, fragments, and antibody conjugates
Catalog No. NC2105715
Add to Cart
Request a Quote