Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Recycling solution for Polypropylene (PP) and Polyethylene (PE) garments
- All brands of disposable apparel are accepted
- Eco-friendly Boxes: Made from recycled materials and easy to assemble
- Collection boxes include a self-locking mechanisms and prepaid shipping label
- Not for recycling of non PP
- Do not recycle biological materials, infectious waste, bloodborne pathogens, hazardous materials or garments exposed to more than 10% chemical exposure of the surface area
- Available in 3 sizes: Small 12 x 12 x 20 in., Medium 12 x 11 x 38 in., Large 15 x 13 x 38 in.
- 5 boxes per case
- Quarterly report available upon request: Reports to include, but not limited to, materials recycled, amount processed, and materials end-of-life scenarios.
Specifications
Specifications
| Capacity (Metric) | 121.43 L |
| Capacity (English) | 26.71 gal. |
| Dimensions (L x W x H) | 15 x 13 x 38 in. |
| Size | Large |
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | Apparel |
| Material | Recycled Materials |
| Product Type | Recycling Box |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
