Learn More
Ubpbio APPBP1/6xHis-UBA3, Human 100 µg
Catalog No. NC2874855
Encompass_Preferred
APPBP1/UBA3 is the heterodimeric E1 enzyme that conjugates Nedd8 to protein substrates. Nedd8 is conjugated onto the catalytic cysteine residue of UBA3 by formation of a thioester bond in an ATP-dependent reaction. The activated Nedd8 is then passed on to an E2 (Ubc12) to be finally attached to a substrate. Nedd8 is mainly conjugated on Cullins to increase the activity of the Cullin – RING E3 Ub ligases.
Catalog No. NC2874855 Supplier Ubpbio Supplier No. B1401
APPBP1/UBA3 is the heterodimeric E1 enzyme that conjugates Nedd8 to protein substrates. Nedd8 is conjugated onto the catalytic cysteine residue of UBA3 by formation of a thioester bond in an ATP-dependent reaction. The activated Nedd8 is then passed on to an E2 (Ubc12) to be finally attached to a substrate. Nedd8 is mainly conjugated on Cullins to increase the activity of the Cullin – RING E3 Ub ligases.