Learn More
Aviva Systems Biology Corp APPBP2 Antibody - middle region (ARP52171_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52171_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APPBP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human APPBP2. Alternate names: HS.84084, KIAA0228, PAT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53683