Learn More
Aviva Systems Biology Corp APPD Antibody - C-terminal region (AVARP09055_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP09055_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against APPD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human APPD. Alternate names: APPD, PHAFIN1, ZFYVE15ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rat, Yeast
Catalog No. 50-014-62591