Learn More
Crescent Chemical Co Inc APPDX IX 8240/8260 6COMP 1ML
SDP
Catalog No. 5067793
Encompass_Preferred
CLP Volatiles Calibration Mix #2 - Gases, 6 Components Bromomethane, Chloroethane, Chloromethane, Dichlorodifluoromethane, Trichlorofluoromethane, Vinyl chloride CAS Size - 1mL Catalog Number - CC2006
Catalog No. 50-677-93 Supplier Crescent Chemical Co Inc Supplier No. CTCC2006
CLP Volatiles Calibration Mix #2 - Gases, 6 Components Bromomethane, Chloroethane, Chloromethane, Dichlorodifluoromethane, Trichlorofluoromethane, Vinyl chloride CAS Size - 1mL Catalog Number - CC2006