Learn More
SPECIALIZEDRX APPLE /FUJI RED/NA POWDER 100G
Catalog No. NC3863024
Encompass
A natural and artificial, anhydrous, water soluble/miscible powder flavor concentrate.Contains no allergensDye-free and gluten-freeEasily incorporates into hard lozenge/candy bases, troche bases, rapid dissolve tablets or oral suspensions.Free-flowingRelatively heat tolerantSuggested concentration range: 1-2 percentTaste and aromaVegan compliant /contains no animal products/
Catalog No. NC3863024 Supplier SPECIALIZEDRX Supplier No. F015540
A natural and artificial, anhydrous, water soluble/miscible powder flavor concentrate.Contains no allergensDye-free and gluten-freeEasily incorporates into hard lozenge/candy bases, troche bases, rapid dissolve tablets or oral suspensions.Free-flowingRelatively heat tolerantSuggested concentration range: 1-2 percentTaste and aromaVegan compliant /contains no animal products/