Learn More
Med Vet International Swabstick Micro-Tip Cotton Tip Wood Shaft 6 Inch NonSterile, A28PC, 200/Box
Catalog No. 503044379
Encompass_Preferred
Applicators have a nonsterile precision miniature tip, only 2.8 millimeters in diameter Swabsticks have a 6-inch birch shaft There are 200 cotton tipped swabs in a box
Catalog No. 50-304-4379 Supplier Med Vet International Supplier No. 555951BX
Applicators have a nonsterile precision miniature tip, only 2.8 millimeters in diameter Swabsticks have a 6-inch birch shaft There are 200 cotton tipped swabs in a box