Learn More
Med Vet International Skin Barrier Wipe Skincote™ 70% Strength Isopropyl Alcohol Individual Packet NonSterile, 1506, 1000/Case
Catalog No. 503046639
Encompass_Preferred
Applied to skin with pre-moistened applicator Provides protective coating on skin When dry, apply bandage or appliance Adhesive or appliance adheres to Skincote dressing, not skin Does not have to be removed when changing bandage or appliance
Catalog No. 50-304-6639 Supplier Med Vet International Supplier No. 246020CS
Applied to skin with pre-moistened applicator Provides protective coating on skin When dry, apply bandage or appliance Adhesive or appliance adheres to Skincote dressing, not skin Does not have to be removed when changing bandage or appliance