Learn More
Hach Company Application Software Brewery Analysis
Supplier:  Hach Company LZV942
Encompass
The application software LZV942 contains 12 brewery assays that conveniently upload to a DR6000 via USB. Assays are based on published and observed brewing methods according MEBAK and include procedures and calculations for: Anthocyanogens, Iron, Steam volatile phenols, Bitter units, Photometric iodine, Thiobarbituric acid number (TAN), Free amino nitrogen, Reductones, Vicinal diketones.
Catalog No. 50-199-7957