Learn More
Hach Company Application Software Drinking Water Analysis
Supplier:  Hach Company LZV941
Encompass
The application software drinking water is a compilation of all methods that are relevant for drinking water analysis. This software allows for individual parameters to be executed without reagent addition through self absorption at various wavelengths. Nitrate (self-absorption), Organic load (SAC), Coloration / organic load (SAC)
Catalog No. 50-199-7956