Learn More
Ace Glass, Inc. Tapered plastic sleeve for use in fitting O-rings on stopcock plugs, size 0-15
SDP
Catalog No. 50974392
Encompass_Preferred
APPLICATOR 0-15 (GLASSBLOWER)
Catalog No. 50-974-392 Supplier Ace Glass, Inc. Supplier No. 819466
APPLICATOR 0-15 (GLASSBLOWER)