Learn More
Ace Glass, Inc. Tapered plastic sleeve for use in fitting O-rings on stopcock plugs, sizes 0-5 & 0-8
SDP
Catalog No. 50979713
Encompass_Preferred
APPLICATOR 0-5 (GLASSBLOWER)
Catalog No. 50-979-713 Supplier Ace Glass, Inc. Supplier No. 819462
APPLICATOR 0-5 (GLASSBLOWER)