Learn More
Med Vet International McKesson Cotton Tip Wood Shaft Swabsticks, 6-Inch Length, 24-106-2S, 2/Pack
Catalog No. 503044556
Encompass_Preferred
Swabstick McKesson Cotton Tip Wood Shaft 6 Inch Sterile 2 per Pack McKesson cotton-tipped applicators have a wooden shaft for a secure, comfortable grip The latex-free absorbent cotton tip holds solutions such as iodine and other disinfectants for application over skin areas The 6-inch cotton swab applicators come individually wrapped to keep them sterile Each wrapper features a ruler for measuring the site of application and gauging the amount of solution needed The medical swabsticks prevent cross-contamination
Catalog No. 50-304-4556 Supplier Med Vet International Supplier No. 508713PK
Swabstick McKesson Cotton Tip Wood Shaft 6 Inch Sterile 2 per Pack McKesson cotton-tipped applicators have a wooden shaft for a secure, comfortable grip The latex-free absorbent cotton tip holds solutions such as iodine and other disinfectants for application over skin areas The 6-inch cotton swab applicators come individually wrapped to keep them sterile Each wrapper features a ruler for measuring the site of application and gauging the amount of solution needed The medical swabsticks prevent cross-contamination