Learn More
Med Vet International McKesson Nonsterile Cotton-Tip Swabsticks, 3 Inch, 24-103, 1000/Box
Catalog No. 503043947
Encompass_Preferred
Swabstick McKesson Cotton Tip Wood Shaft 3 Inch NonSterile 100 per Pack These McKesson cotton-tipped applicators are ideal for applying ointments, cleaning wounds and collecting specimens They have soft, bulb-shaped cotton tips to enhance comfort during application These non-sterile swabsticks are absorbent They feature wooden shafts that are sturdy, versatile and biodegradable These cotton swabsticks are latex-free to prevent irritation
Catalog No. 50-304-3947 Supplier Med Vet International Supplier No. 508715BX
Swabstick McKesson Cotton Tip Wood Shaft 3 Inch NonSterile 100 per Pack These McKesson cotton-tipped applicators are ideal for applying ointments, cleaning wounds and collecting specimens They have soft, bulb-shaped cotton tips to enhance comfort during application These non-sterile swabsticks are absorbent They feature wooden shafts that are sturdy, versatile and biodegradable These cotton swabsticks are latex-free to prevent irritation