Learn More
Med Vet International McKesson Swabsticks, Cotton-Tip, Wood Shaft, 6 Inch, 24-106, 10000/Case
Catalog No. 503044663
Encompass_Preferred
Swabstick McKesson Cotton Tip Wood Shaft 6 Inch NonSterile 100 per Pack These McKesson cotton-tipped applicators are ideal for applying ointments, cleaning wounds and collecting specimens They have soft, bulb-shaped cotton tips to enhance comfort during application These non-sterile swabsticks are absorbent They feature wooden shafts that are sturdy, versatile and biodegradable These 6-inch swabs are latex-free to prevent irritation
Catalog No. 50-304-4663 Supplier Med Vet International Supplier No. 508716CS
Swabstick McKesson Cotton Tip Wood Shaft 6 Inch NonSterile 100 per Pack These McKesson cotton-tipped applicators are ideal for applying ointments, cleaning wounds and collecting specimens They have soft, bulb-shaped cotton tips to enhance comfort during application These non-sterile swabsticks are absorbent They feature wooden shafts that are sturdy, versatile and biodegradable These 6-inch swabs are latex-free to prevent irritation