Learn More
Med Vet International Dynarex Skincote ProtectIVe Dressing Applicator, 1000/Case
Catalog No. 502829868
Encompass_Preferred
Skincote Protective Dressing Applicator protects skin against irritation from adhesives, appliances, electrodes, and external catheters. Designed to provide a protective barrier between adhesives, tapes and skin Applies to skin with pre-moistened foil packed applicator Adhesives and ostomy appliances adhere to protective plastic layer, not skin, for optimal comfort Does not need to be removed when changing bandages or appliances
Catalog No. 50-282-9868 Supplier Med Vet International Supplier No. 1506
Skincote Protective Dressing Applicator protects skin against irritation from adhesives, appliances, electrodes, and external catheters. Designed to provide a protective barrier between adhesives, tapes and skin Applies to skin with pre-moistened foil packed applicator Adhesives and ostomy appliances adhere to protective plastic layer, not skin, for optimal comfort Does not need to be removed when changing bandages or appliances