Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ DNA Extract All Reagents Kit

Catalog No. 4403319
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
20 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-033-19 5 mL
44-026-16 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44-033-19 Supplier Applied Biosystems™ Supplier No. 4403319
  • $246.00
    $223.65 / Each of 1

    Save $22.35

    9% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Provides PCR-ready DNA from a wide variety of sample types, ranging from blood to buccal swabs to plant tissues, in 5 minutes

Rapid sample prep for qPCR genotyping

Rapid DNA extraction kit for qPCR SNP genotyping

DNA Extract All™ Reagents Kit is a rapid DNA extraction kit designed for preparation of PCR-ready crude lysates for qPCR-based SNP genotyping. The kit enables fast, simple sample preparation from a wide range of biological sample types—including buccal swabs, blood, tissue, cultured cells, and plant material—in as little as 5 minutes, without column- or magnetic bead–based DNA purification.

This streamlined, column-free and bead-free workflow produces crude lysates suitable for downstream singleplex and multiplex SNP genotyping, eliminating traditional wash and elution steps associated with genomic DNA purification.

Compared to column-based and magnetic bead–based purification methods, DNA Extract All reduces workflow complexity with up to 85% fewer steps, minimizing hands-on time and simplifying high-throughput genotyping workflows.

Simple, three-step sample preparation workflow

  1. Mix sample with the provided DNA lysis solution
  2. Incubate for 3 minutes
  3. Add the DNA stabilizing solution

The resulting crude lysate is ready for qPCR and supports both Fast and Standard thermal cycling conditions, depending on the selected master mix.

For a typical batch of 48 samples, DNA Extract All requires approximately half the processing time compared to conventional DNA purification workflows, enabling faster turnaround from sample to SNP genotyping results.

Validated performance for multiplex SNP genotyping

DNA Extract All–generated crude lysates have been evaluated for multiplex qPCR SNP genotyping using TaqPath™ ProAmp™ Master Mix.

In internal testing using selected sets of 2-SNP multiplex assay configurations and 3-SNP multiplex configurations*, crude lysates prepared from buccal swabs and blood demonstrated 100% concordance with genotype calls obtained from purified genomic DNA**.

When used together with TaqPath™ ProAmp™ Master Mix, the DNA Extract All stabilizing solution helps condition crude lysates and reduce the impact of PCR inhibitors commonly present in biological samples. This combined workflow supports robust amplification and accurate SNP genotype calls without traditional DNA purification.

These data demonstrate that DNA Extract All™ Reagents Kit enables reliable multiplex SNP genotyping performance while significantly reducing sample preparation time and complexity.

Designed for SNP genotyping workflows (not CNV analysis)

DNA Extract All crude lysates are intended for SNP genotyping applications.

For copy number variation (CNV) analysis, purified genomic DNA is required. Crude lysate inputs are not recommended for CNV applications to ensure accurate copy number determination.

Part of a complete Sample-to-Genotype workflow

DNA Extract All™ Reagents Kit integrates into a complete Sample-to-Genotype workflow, including:

  • TaqMan™ Predesigned SNP Genotyping Assays or TaqMan™ DME Genotyping Assays
  • TaqPath™ ProAmp™ Master Mix for singleplex and multiplex SNP genotyping
  • QuantStudio™ Real-Time PCR Instruments
  • Diomni™ Design and Analysis Software for multiplex genotyping data analysis

This end-to-end workflow supports high-throughput SNP genotyping with reduced sample preparation time and consistent genotype accuracy.

Notes

  • Thermal cycling conditions should follow the recommendations for the selected master mix.
  • *3-SNP assay multiplexing requires a custom version of TaqPath™ ProAmp™ Master Mix without ROX.
  • **Performance claims are based on internal testing using selected sets of multiplex TaqMan™ Predesigned SNP Genotyping Assays or TaqMan™ DME Genotyping Assays, comparing DNA Extract All crude sample lysates to purified DNA.

Recommended Storage

Store at 2° to 8°C.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2–8°C.
Product Line Extract All
Quantity 5 mL
Reagent Type Cell Lysis Buffer
Test Time 5 min
Form Liquid
Is there a specific protocol for preparing the lysate to be used with TaqPath ProAmp Mastermix (Cat. No. A30865)?

The standard protocols described in the user guides of the DNA Extract All Reagent Kit and MagMAX DNA Multi-Sample Ultra Kit should be followed.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.