Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ FLoid™ Printer Ink/Paper

Catalog No. 4472389
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Floid Cell Imaging System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
44-723-89 Floid Cell Imaging System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-723-89 Supplier Applied Biosystems™ Supplier No. 4472389
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Color ink and paper set. 18 prints per pack.

For use with FLoid Cell Imaging Station-compatible printer

Print beautiful, borderless, credit-card- sized photos in vibrant colors. The sticker-based backing allows to paste directly into lab notebook.

  • All-in-one color ink cartridge and labels compatible with the FLoid printer
  • Color ink and label set produces credit-card-sized label
  • Adhesive back can be attached to lab notebook
  • Water-resistant prints last up to 100 years

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Printer Ink/Paper
For Use With (Equipment) Floid Cell Imaging System
Includes Color Ink, Paper Set
Quantity 1 pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.