Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ GeneAmp™ Thermal Isolation Frame

Catalog No. 4308927
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 frames
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-089-27 8 frames
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-089-27 Supplier Applied Biosystems™ Supplier No. 4308927
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

8 GeneAmp Thermal Isolation Frames.

Balances thermal uniformity across 0.5mL GeneAmp PCR System 9700

When used with the 0.5mL GeneAmp reaction tubes, the GeneAmp Thermal Isolation Frame creates a proper thermal and physical seal between the heated cover and the reaction tubes.

  • Sturdy,reinforced fiberglass ensures precise PCR results
  • Convenient tool protects precious samples
  • Maintain Sample Temperature Uniformity
  • During PCR, it is critical that thermal cycler lid does not compress sample tubes and heat samples capriciously
  • Fits snugly around samples to ensure reproducible results
  • Designed for Convenience
  • Fits around thermal cycler sample block
  • When thermal cycler lid is closed, frame ensures even contact between lid and tubes
  • Leave frame on thermal cycler for continual back-to-back runs

See user's manual or package insert for limited label license and trademark information.

PCR, PCR and Real-Time PCR

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 8 GeneAmp™ Thermal Isolation Frames. Store at room temperature.
Product Line GeneAmp
Color Black
Product Type Thermal Isolation Frame
For Use With (Equipment) GeneAmp 9700
For Use With (Application) PCR
Quantity 8 frames
Shipping Condition Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.