Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MagMAX™ DNA Multi-Sample Ultra Kit (with extra proteinase K and DNA binding beads)

Catalog No. p-4926393
Encompass_Preferred
Change view
Click to view available options
Quantity:
2,500 preps
500 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25919 500 Preps
A25920 2,500 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25919 Supplier Applied Biosystems™ Supplier No. A25919
Only null left
Add to Cart
Add to Cart

The MagMAX™ DNA Multi-Sample Ultra Kit is optimized to isolate genomic DNA (gDNA) from a variety of samples, such as whole blood, buccal cells, saliva, urine, blood cards, mouth rinse, and tissue.

Note: This product is identical to the MagMAX™ DNA Multi-Sample Ultra Kit (Cat. No. A25597) except that it comes with additional proteinase K and DNA Binding Beads, which some customers find useful for their specific applications.

The MagMAX™ DNA Multi-Sample Ultra Kit is optimized to isolate genomic DNA (gDNA) from a variety of samples, such as whole blood, buccal cells, saliva, urine, blood cards, mouth rinse, and tissue. gDNA purified with the kit is ideal for qPCR applications on sensitive, scalable platforms such as the QuantStudio™ 12K Flex System with OpenArray™ Block (with AccuFill™ System). The magnetic bead–based purification format allows you to easily scale from processing 12 to 500 samples a day, making it the ideal choice for pharmacogenomics (PGx) studies.

Features of the MagMAX™ DNA Multi-Sample Ultra Kit include:
• Streamlined protocols for numerous non-invasive biological samples
• Optimized to provide DNA yields suitable for OpenArray™ and Ion AmpliSeq™ applications
• Packaged for high-throughput needs

One Kit for a Diverse Set of Sample Types
Why purchase multiple purification kits to isolate gDNA from different sample types? The MagMAX™ DNA Multi-Sample Ultra Kit eliminates the headache of ordering these extra kits because it has been optimized for use with a variety of biological samples. The kit is designed for high-throughput isolation of gDNA from 1 buccal swab, 50 to 350 μL of whole blood, up to 400 μL urine (without sample concentration), up to 150 μL saliva, 1-5 mL mouth rinse, blood spot cards, and mammalian tissue samples (up to 10 mg). The protocols and magnetic bead format make the kit fully amenable to automation.

*Please note that the Thermo Scientific™ KingFisher™ Flex Magnetic Particle Processor with a 24 Deep-well Head is recommended for sample volumes greater than 50 μL.

Benefits of MagMAX™ Magnetic Bead–based Purification
Magnetic beads offer many benefits compared to other technologies for isolating gDNA. Beads bind gDNA more efficiently than glass fiber filters, resulting in higher and more consistent gDNA yields. Additionally, because filters are not used, there is no risk of filter clogging due to cellular particulates in samples.

Accessory Products
Proteinase K (Cat. No. A25561), DNA Binding Beads (Cat. No. A25562), Wash Solution 1 Concentrate (Cat. No. AM8504), and Wash Solution 2 Concentrate (Cat. No. AM8640) are components of the kit that may also be purchased separately.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • DNA Binding Beads, 2 x 8 mL (store at 2°C to 8°C, DO NOT FREEZE)
• DNA Elution Buffer 1, 1 x 25 mL (store at room temperature)
• DNA Elution Buffer 2, 1 x 25 mL (store at room temperature)
• Wash Solution 1 Concentrate, 1 x 80 mL* (store at room temperature)
• Wash Solution 2 Concentrate, 1 x 162 mL* (store at room temperature)
• Multi-Sample DNA Lysis Buffer, 1 x 100 mL (store at room temperature)
• Nuclease-Free Water, 1 x 100 mL (store at room temperature)
• PK Buffer, 1 x 96 mL (store at room temperature)
• Proteinase K, 2 x 4 mL (store at -5° to -30°C)
• Ribonuclease A (RNase A), 1 x 1.25 mL (store at -5° to -30°C)
*after addition of alcohol
Format Kit
Sample Type Blood, Buccal Samples, Cells, Liquid Samples (e.g. Serum), Tissue
For Use With (Application) Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR)
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
No. of Reactions 500 Preps
Product Line MagMAX
Product Type DNA Multi-Sample Ultra Kit
Quantity 500 Preps
Target Genomic DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.