Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ 96-Well Full Plate Cover

Catalog No. N8010550
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 covers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N8010550 5 covers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N8010550 Supplier Applied Biosystems™ Supplier No. N8010550
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Seal up to 96-wells in MicroAmp Tray/Retainer Set or Reaction Plates to prevent evaporation during thermal cycling of samples

  • Durable design allows for continuous use with high-quality results
  • Convenient, cost-saving alternative to MicroAmp Caps
  • Patented one-piece design for convenience
  • Reusable up to five times
  • Manufactured from PCR-compatible rubber
  • Non-sterile and autoclavable
  • Platform: GeneAmp™ 9700

See user's manual or package insert for limited label license, and trademark information.

PCR, PCR & Real-Time PCR

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Content And Storage MicroAmp™ 96-Well Full Plate Cover is manufactured from PCR-compatible rubber. It is non-sterile, autoclavable and guaranteed reusable up to five times.
Packaging 5 Covers
Product Line MicroAmp
Color Natural
Product Type Microplate Lid
For Use With (Equipment) 2720 Thermal Cycler, 6100 Thermal Cycler, GeneAmp 9700, 2720 Thermal Cycler
No. of Wells 96
Sterility Non-sterile
For Use With (Application) PCR
Plate Compatibility 96-Well Plates
Quantity 5 covers
Shipping Condition Room Temperature
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.