Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ 96-Well Tray for VeriFlex™ Blocks

Catalog No. 4379983
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Trays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-799-83 10 Trays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-799-83 Supplier Applied Biosystems™ Supplier No. 4379983
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

10 MicroAmp 96-Well Trays for VeriFlex Blocks.

Engineered for efficient sample processing

• Engineered for efficient sample processing
• Designed specifically for MicroAmp™ Fast Reaction Tubes with Caps (0.1 mL) and MicroAmp™ Fast 8-Tube Strips (0.1 mL)
• Designed specifically for MicroAmp™ Reaction Tubes with Caps (0.2 mL)
• Manufactured to high specification, ensuring a quality product

Convenience and Performance The MicroAmp™ 96-Well Tray for VeriFlex™ Blocks allows you to organize your tubes while adding sample and mix.This handy tray reduces spillage and tube mix-ups by securing and ordering your tubes on the lab bench. When the samples are ready, simply put the tray of tubes directly on the block of the StepOne™ Real-Time PCR System or Veriti™ Thermal Cycler. Not only does the tray eliminate the hassle of transferring the tubes, but it also provides the thermal contact necessary for thermal cycling.

Note: See user's manual or package insert for limited label license and trademark information. For Research Use Only. Not for use in diagnostics procedures.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 10 MicroAmp™ 96-Well Trays for VeriFlex™ Blocks. Store at room temperature.
Product Line MicroAmp
Color Black
Product Type Tray
For Use With (Equipment) 2720 Thermal Cycler, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ProFlex™ PCR System
For Use With (Application) PCR
Plate Compatibility 96-Well Plates
Quantity 10 Trays
Shipping Condition Room Temperature
Why are my PCR tubes crushed/deformed after my PCR run?

The heated thermal cycler lid is designed to make sure that optimized pressure is applied on the microplate for an efficient PCR reaction. When using PCR tubes, excess pressure on the tubes, from the heated lid can cause deformation of the tubes. To avoid this, we recommend using the Tray/Retainer set, as shown below:

Cat. No. 4381850, MicroAmp 96-Well Tray/Retainer Set for Veriti Systems – This Tray is for use with MicroAmp Strip Tubes (or MicroAmp Reaction Tubes without Caps, 0.2 mL), with the Veriti Thermal Cycler, SimpliAmp Thermal Cycler, and Proflex PCR System.
Cat. No. 403081, MicroAmp 96-Well Tray/Retainer Set – This Tray is for use with MicroAmp Strip Tubes (or MicroAmp Reaction Tubes without Caps, 0.2 mL), with the 7000 System, 2720 Thermal Cycler, and GeneAmp PCR System 9700.
Cat. No. 4379983, MicroAmp 96-Well Tray for VeriFlex Blocks – This Tray is for use with MicroAmp Strip Tubes (or MicroAmp Reaction Tubes with Caps, 0.1 mL), with the Veriti Thermal Cycler, Veriti Fast Thermal Cycler, SimpliAmp Thermal Cycler, and Proflex PCR System.

Please take a look at this video: How to Use Adapter Tray Retainers in Thermal Cycler Starter Kits (https://www.youtube.com/watch?v=1uh2ZxZJtgs).

Why does the SimpliAmp Thermal Cycler starter kit contain two different types of trays?

The two different types of trays are for use with different tubes:

-MicroAmp 96-well Tray/Retainer Set for Veriti Systems (Cat. No. 4381850) - this tray is for use with 0.2 mL single tubes without caps (Cat. Nos. N8010533, N8011533), 0.2 mL 8-tube strip with attached dome caps (Cat. No. A30589), and 0.2 mL tube strips without caps (Cat. No. 4316567)
-MicroAmp 96-well Tray for VeriFlex Blocks (Cat. No. 4379983) - this tray is for use with 0.2 mL single tubes with caps (Cat. Nos. N8010540, N8011540)

Can I run strip tubes on the ViiA 7 instrument?

Yes. Make sure to use the appropriate matching parts. For the 96-well standard block, use the standard 96-well plate adapter (Cat. No. 4459845), the retainer tray for (0.2 mL) tubes, and Optical 8-Tube Strips and Caps (Cat. No. 4316567 and 4323032). For the 96-well Fast block, use the Fast 96-well plate adapter (Cat. No. 4462078), the retainer tray (Cat. No. 4379983), and Fast (0.1 mL) Optical 8-tube Caps and Strips (Cat. Nos. 4323032 and 4358293).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.