Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Fast Red Reaction Plates with Barcode

Catalog No. 4483492
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-834-92 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-834-92 Supplier Applied Biosystems™ Supplier No. 4483492
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments

  • Optimized for performance with Applied Biosystems instruments
  • Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
  • Constructed to ANSI™/SBS standard, well suited to high-throughput laboratories and robot handling
  • Great mechanical stability and flatness for complete workflow needs
  • Use of polypropylene and high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate provides mechanical stability, flatness, and excellent durability
  • Ideal choice for use with automation/robotics and multi-instrument experiments (Like thermal cycler, real-time thermal cycler, genetic analyzer)
  • Thin-wall construction allows for snug fit to thermal blocks with excellent heat transfer from thermal block to sample
  • Easy-to-read, black well identification text helps to minimize pipetting mistakes and improves sample preparation productivity

  • Platform: 310 Genetic Analyzer, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7500 Fast Dx System, 7500 Fast System, 7900HT Fast System, StepOnePlus™, Veriti™ Dx Fast Thermal Cycler, Veriti Fast Thermal Cycler, ViiA™ 7 Dx Fast System, ViiA 7 Fast System

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Certifications/Compliance Certified DNA/RNase Free
Content And Storage Store at room temperature.
Packaging 20 Plates
Product Line MicroAmp
Color Optical, Red
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Semi
Material Polycarbonate, Polypropylene
Volume (Metric) Well 0.2 mL
No. of Wells 96
Volume (Metric) Working 0.1 mL
Quantity 20 Plates
Reaction Speed Fast
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.