Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ Fast 48-Well Tray

Catalog No. 4375282
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Trays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-752-82 10 Trays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-752-82 Supplier Applied Biosystems™ Supplier No. 4375282
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

10 MicroAmp™ Fast 48-Well Trays

Engineered for efficient sample processing

Specifically designed to snugly hold MicroAmp™ Fast Reaction Tubes with Cap (0.1mL) or the MicroAmp™ Fast 8-Tube Strip (0.1mL), is required for more efficient sample processing on the Applied Biosystems™ StepOne™ Real-Time PCR System.

  • Manufactured to high specification, ensuring quality product
  • Allows you to organize tubes while adding sample and mix
  • Handy tray reduces spillage and tube mix-ups by securing and ordering tubes on lab bench
  • When samples are ready, simply put the tray of tubes directly on the block of StepOne™ Real-Time PCR System
  • Tray eliminats hassle of transferring tubes and also provides thermal contact necessary for thermal cycling
Note:See user's manual or package insert for limited label license and trademark information.

For Research Use Only. Not for use in diagnostic procedures.

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 10 MicroAmp™ Fast 48-Well Trays. Store at room temperature.
Packaging 10 Trays
Product Line MicroAmp
Product Type 48-Well Tray
For Use With (Equipment) StepOne™
No. of Wells 48
For Use With (Application) PCR
Plate Compatibility 48-Well Plates
Quantity 10 Trays
Shipping Condition Room Temperature
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.