Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ Fast 96-Well Tray

Catalog No. 4358305
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Trays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-583-05 10 Trays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-583-05 Supplier Applied Biosystems™ Supplier No. 4358305
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

10 MicroAmp Fast 96-Well Trays.

Designed to snugly hold MicroAmp Fast reaction tubes with cap (0.1mL) and MicroAmp Fast 8-tube strips

The MicroAmp fast 96-well tray is required for more efficient sample processing on the Applied Biosystems9800 Fast Thermal Cycler.

  • Organizes fast reaction tubes or strips
    Provides thermal contact necessary for fast PCR
    Manufactured to high specification, ensuring quality production

  • Allows organize tubes while adding sample and mix
  • Reduces spillage and tube mix-ups by securing and ordering tubes on lab bench
  • When samples are ready, tray of tubes are put directly on block of 9800 Fast Thermal Cycler
  • Eliminates hassle of transferring tubes and provides thermal contact necessary for efficient and accurate thermal cycling

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 10 MicroAmp™ Fast 96-Well Trays. Store at room temperature.
Product Line MicroAmp
Product Type Tray
For Use With (Equipment) 9800 Fast Thermal Cycler
For Use With (Application) PCR
Plate Compatibility 96-Well Plates
Quantity 10 Trays
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.