Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ Fast Optical 48-Well Reaction Plate

Catalog No. 4375816
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-758-16 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-758-16 Supplier Applied Biosystems™ Supplier No. 4375816
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

20 plates

Optimized to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification

  • Constructed from single rigid piece of polypropylene in 48-well format
  • Compatible with StepOne™ Real-Time PCR System
  • Maximum thermal conductivity for precise thermal cycling
  • Reproducible, specific, and sensitive real-time PCR results in just 40 minutes
  • Validated with other system components for consistent results
  • Each reaction plate includes unique serialized, eight character number label (barcode) that is user-readable and machine-readable to prevent tracking errors
  • Advanced design dramatically increases thermal contact with StepOne Real-Time PCR System
  • Consistent well-to-well thickness helps samples cycle through PCR process uniformly to reduce experimental artifacts and help ensure fast, accurate results

For Research Use Only. Not for use in diagnostic procedures.

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 20 plates.

May be stored at room temperature. Under proper storage conditions, plates may be stored indefinitely.
Packaging 20 Plates
Product Line MicroAmp
Color Optical
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Semi
Material Polypropylene
Volume (Metric) Well 0.2 mL
No. of Wells 48
Volume (Metric) Working 0.1 mL
Quantity 20 Plates
Reaction Speed Fast
Shipping Condition Room Temperature
Show More Show Less
Can I cut a 96-well FAST plate to fit the StepOne system?

No, the MicroAmp Fast Optical 48-Well Reaction Plate (P/N 4375816) is the only plate that can be used on the StepOne system. Using a plate that has not been designed for use with the StepOne Real-Time PCR System is not recommended and can potentially damage the instrument.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.