Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ Multi Removal Tool

Catalog No. 4313950
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Tool
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-139-50 1 Tool
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-139-50 Supplier Applied Biosystems™ Supplier No. 4313950
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

1 MicroAmp Multi Removal Tool

Easily removes caps from tubes, microplates from thermal cyclers and more

This multi-functional tool makes it easier to remove caps from tubes, microplates from thermal cyclers, and much more. Use to open MicroAmp tubes or GeneAmp tubes with attached caps, remove microplates from a thermal cycler sample block, and remove strips of caps from reaction tubes in a microplate and Tray/Retainer assembly.

  • Multi-edged tool streamlines repetitive tasks
  • Easy-to-use tool makes frustrating steps effortless
  • Sturdy construction extends product life
  • Versatile and Ergonomic
  • Multiple edges with design features that facilitate separation of caps from tubes, microplates from thermal cyclers, tubes from thermal cyclers, and tubes from microplates
  • One edge contains U-shaped notch to open tube caps
  • One edge has ridged tongue to remove strip of capped tubes from microplate
  • Two edges have prongs to remove microplates from thermal cyclers
  • Two hook protrusions can insert into short side of microplate and lift it from sample block
  • Quick and Easy tool speeds up routine steps of large experiment and reduces dropped plates and spilled samples
    • See user's manual or package insert for limited label license, and trademark information.

      PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 1 MicroAmp™ Multi Removal Tool. Store at room temperature.
Product Line MicroAmp
Product Type Multi Removal Tool
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
For Use With (Application) PCR
Quantity 1 Tool
Shipping Condition Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.