Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ Optical 96-Well Reaction Plate with Barcode

Catalog No. p-4918085
Encompass_Preferred
Change view
Click to view available options
No. of Wells:
96
Quantity:
20 Plates
500 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells Quantity
43-067-37 96 20 Plates
43-266-59 96 500 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-067-37 Supplier Applied Biosystems™ Supplier No. 4306737
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Optimized to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification

  • Constructed from single rigid piece of polypropylene in a 96-well format
  • Compatible with Applied Biosystems 96-Well Real-Time PCR systems and thermal cyclers
  • Frosted to minimize interfering fluorescence from wells of cycling block
  • Screened to eliminate autofluorescent plates
  • Unique design provides barrier to ambient air to help ensure well-to-well temperature uniformity
  • Each reaction plate includes unique serialized, eight character number label (barcode) that is user-readable and machine-readable to prevent tracking errors
  • Capacity: 96 x 0.2mL tubes

  • Platform: 2720 Thermal Cycler, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 6100 Nucleic Acid PrepStation, 7000 System, 7300 System, 7500 System, 7900HT System, GeneAmp™ 9700, ProFlex™ PCR System, QuantStudio™, QuantStudio™ Dx, SimpliAmp™ Thermal Cycler, Veriti™ Dx Thermal Cycler, Veriti™ Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System

  • Applications: PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Ambient

TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 20 plates.

May be stored at room temperature. With proper storage, plates may be stored indefinitely.
Packaging 20 Plates
Product Line MicroAmp
Color Optical
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Semi
Material Polypropylene
Volume (Metric) Well 0.3 mL
No. of Wells 96
Volume (Metric) Working 0.2 mL
Quantity 20 Plates
Reaction Speed Standard
Shipping Condition Room Temperature
Show More Show Less
Are the MicroAmp Optical 96-Well Reaction Plate with Barcode (Cat. No. 4306737, 4326659) and the one without Barcode (Cat. No. N8010560, 4316813) the same product?

Yes, both the plates are the same. The only difference between them is the barcode.

Can I place a standard MicroAmp 96-Well Optical Reaction Plate onto a 7500 Fast Real-Time PCR System?

No. The plate does not fit and will result in the crushing of the wells and possible contamination of the block due to leaking of the well contents.

What does the SeqStudio Starter Kit (PN A35000) contain?

The SeqStudio Starter Kit (PN A35000) includes:
- SeqStudio Integrated Capillary Protector (Cat. No. A31923)
- SeqStudio Cathode Buffer Container (Cat. No. A33401)
- Sequencing Standard, BigDye Terminator v3.1, for 3500/SeqStudio Flex (Cat. No. 4404312)
- DS-33 GeneScan Installation Standards with GeneScan 600 LIZ Size Standard v2.0 (Cat. No. 4376911)
- Hi-Di Formamide (Cat. No. 4401457)
- MicroAmp Optical 96-Well Reaction Plate with Barcode (Cat. No. 4306737)
- MicroAmp 96-Well Tray/Retainer Set (Cat. No. 403081)
- MicroAmp Optical 8-Tube Strip, 0.2 mL (Cat. No. 4316567)
- Nuclease-Free Water (not DEPC-Treated) (Cat. No. AM9930)
- Cathode Buffer Container Reservoir Septa for SeqStudio Genetic Analyzer (Cat. No. A35640)
- 96-well Septa for SeqStudio Genetic Analyzer (Cat. No. A35641)
- 8-strip Septa for SeqStudio Genetic Analyzer (Cat. No. A35643)
- SeqStudio Cartridge v2 (PN A41328) (if this component is ordered online the catalog number is A41331)

Which of your plastic consumables are compatible with the Applied Biosystems 7900HT Fast Real-Time PCR System?

There are many of our reagents and plastic consumables that can be used on the Applied Biosystems 7900HT Fast Real-Time PCR System. Here are a few examples to help you get started:
Cat. No. 4313663, MicroAmp Optical Adhesive Film Kit
Cat. No. 4311971, 4360954, MicroAmp Optical Adhesive Film
Cat. No. 4323032, MicroAmp Optical 8-Cap Strip
Cat. No. 4309849; 4326270, MicroAmp Optical 384-Well Reaction Plate with Barcode
Cat. No. 4306737; 4326659; 4314320, MicroAmp Optical 96-Well Reaction Plate with Barcode
Cat. No. 4346906, MicroAmp Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL
Cat. No. 4312063, Splash Free 96-Well Base

For more comprehensive lists of compatible plastics, enter "Real-Time PCR Plastics" in the main Search field on our website and follow the links.

What is the difference between the MicroAmp Optical Adhesive Covers and the MicroAmp Optical Caps? Can they be used interchangeably?

The MicroAmp Optical Adhesive Covers (P/N 4311971) have a higher optical efficiency than the MicroAmp Optical Caps, (8 caps/strip) (P/N 4323032). They can be used instead of the MicroAmp Optical Caps only when using the MicroAmp 96-Well Reaction Plate with Barcode (P/N 4306737, 4309849) or the MicroAmp Optical 96-Well Reaction Plate (P/N N801-0560). The Optical Adhesive Covers cannot be used with the MicroAmp Optical Tubes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.