Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ Optical 96-Well Reaction Plate with Barcode & Optical Caps

Catalog No. 403012
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-301-2 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-301-2 Supplier Applied Biosystems™ Supplier No. 403012
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

20 plates with 300 Optical 8-cap Strips, (equivalent to catalog No. 4306737 and 4323032)

Optimized to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification

This plate, constructed from a single rigid piece of polypropylene in a 96-well format, is compatible with Applied Biosystems96-Well Real-Time PCR systems and thermal cyclers.

  • Frosted to minimize interfering fluorescence from wells of cycling block
  • Screened to eliminate auto-fluorescent plates
  • Design provides barrier to ambient air to help insure well-to-well temperature uniformity
  • Each reaction plate includes unique serialized, eight character number label (barcode) that is user-readable and machine-readable to prevent tracking errors
  • Includes MicroAmpOptical 8-Cap Strips
  • MicroAmp disposables are optimized to provide unmatched temperature accuracy and uniformity for fast, oil-free PCR amplification
  • Proprietary MicroAmpplates are essential for high-performance PCR and help ensure greatest temperature and amplification uniformity for samples

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 20 plates with 300 Optical 8-cap Strips
(equivalent to catalog # 4306737 and 4323032)
Product Line MicroAmp
Color Optical
Product Type 96-Well Reaction Plate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
No. of Wells 96
For Use With (Application) PCR
Quantity 20 Plates
Reaction Speed Standard
Show More Show Less
Can I place a standard MicroAmp 96-Well Optical Reaction Plate onto a 7500 Fast Real-Time PCR System?

No. The plate does not fit and will result in the crushing of the wells and possible contamination of the block due to leaking of the well contents.

Which well plates do you recommend using with the QuantStudio 5 Real-Time PCR System?

Please see the following link to our qPCR and PCR Plastics Selection Tool to narrow down the list of products compatible with the QuantStudio 5 Real-Time PCR System: https://www.thermofisher.com/de/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html
- For Manufacturer, Select "Thermo Fisher Scientific"
- For Model, Select "Applied Biosystems QuantStudio 5"
- Select the appropriate block format for your qPCR instrument
- Click "Display Results"
To the left of the results list, you can filter the results further by various parameters (i.e., Consumable Type, Number of Wells, etc.)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.