Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ Snap-On Optical Film Compression Pad

Catalog No. 4333292
Encompass_Preferred
Change view
Click to view available options
Quantity:
9 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-332-92 9 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-332-92 Supplier Applied Biosystems™ Supplier No. 4333292
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

9 each. Compliant cover bounded to a metal frame and designed for use on the ABI PRISM 7900HT Sequence Detection System when using the available automation accessory with 96-well plates

Optical film compression rad attached to metal frame

Designed for use with Applied Biosystems 7900HT Fast Real-Time System when using automation accessory with 96-well plates.

PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 9 each. Compliant cover bounded to a metal frame and designed for use on the ABI PRISM™ 7900HT Sequence Detection System when using the available automation accessory with 96-well plates
Product Line MicroAmp
Color Optical
Product Type Compression Pad
For Use With (Equipment) 7900HT System
For Use With (Application) PCR
Plate Compatibility 96-Well Plates
Quantity 9 each
Shipping Condition Room Temperature
Can the MicroAmp Optical Adhesive Film be used with MicroAmp Optical Tubes?

No. The Optical Adhesive Film is recommended for use only with the MicroAmp Optical 96-well Reaction Plate (with or without barcode) and MicroAmp 384-well Optical Reaction Plate.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.