Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Applied Biosystems™ QuantStudio™ 5 Strip Plate Tray

Catalog No. A55332 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55332 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A55332 Supplier Thermo Scientific™ Supplier No. A55332
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Setup, transfer and run the SureTect PCR assays in the QuantStudio 5 instrument using the QuantStudio 5 Strip Plate tray.

Setup, transfer and run the Thermo Scientific™ SureTect™ PCR Assays in the Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System using the Applied Biosystems™ QuantStudio™ 5 Strip Plate Tray.

Designed specifically for use with the Thermo Scientific™ SureTect™ PCR Assay workflow the QuantStudio 5 Strip Plate tray is used alongside the Thermo Scientific™ 96-well PCR Base to easily setup and transfer PCR tubes into the QuantStudio 5 instrument regardless of sample throughput enabling a simple, streamlined workflow for maximum efficiency and productivity.

Use the QuantStudio 5 Strip Plate Tray to setup, transfer and run your SureTect Assays in the Quant Studio 5 instrument.
 

Designed to work in conjunction with the Thermo Scientific 96 Well PCR Base, the QuantStudio 5 Strip Plate Tray enables you to streamline your SureTect workflow regardless of sample throughput.
 

Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.
 

We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Therrmo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.

TRUSTED_SUSTAINABILITY

Specifications

Sample Type Food
Description Applied Biosystems™ QuantStudio™ 5 Strip Plate Tray
Content And Storage Room Temperature
For Use With (Equipment) 0.1 mL block Quant Studio 5 Instrument
Quantity 10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.