Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Applied Biosystems™ SimpliAmp™ Strip Plate Tray

Catalog No. A55333 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55333 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A55333 Supplier Thermo Scientific™ Supplier No. A55333
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ease of sample set up and reduced handling steps enabled using the SimpliAmp Strip Plate Tray.

Setup and transfer the SureTect lysis tubes into the SimpliAmp instrument using the Applied Biosystems™ SimpliAmp™ Strip Plate Tray.

Designed specifically for use with the Thermo Scientific™ SureTect™ PCR Assay workflow the SimpliAmp Strip Plate tray is used alongside the Thermo Scientific™ 96 Well Base to easily setup and transfer lysis tubes into the SimpliAmp instrument, regardless of sample throughput, reducing hands on steps and enabling a simple, streamlined workflow for maximum efficiency and productivity in your laboratory.

Use the SimpliAmp Strip Plate Tray to easily setup and transfer your SureTect lysis tubes into the SimpliAmp instrument.

Designed to work in conjunction with the Thermo Scientific 96 well base, the SimpliAmp Strip Plate Tray enables you to streamline your Thermo Scientific™ SureTect™ PCR Assay Workflow regardless of sample throughput.

Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.

We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.

TRUSTED_SUSTAINABILITY

Specifications

Sample Type Food
Description Applied Biosystems™ SimpliAmp™ Strip Plate Tray
For Use With (Application) Food safety
For Use With (Equipment) Thermo Scientific™ SureTect™ PCR Assay workflow
Product Line SureTect
Quantity 10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.