Learn More
Applied Scientific Pharmaceutical Coil
SDP
Catalog No. 1264041
Encompass_Preferred
Catalog No. 12-640-41 Supplier Applied Scientific Supplier No. AS28843
Pull apart or cut to any length to plug any size/diameter vial opening or bottle
Coil packed materials will absorb 22 times their weight in moisture.
Rayon Rope
  • Uniform, man-made fiber (cellulose) Recommended for cotton-tearing equipment
Cotton Coil
  • Natural fiber
  • Reoxygenates faster in bottle, resulting in a drier environment
Specifications
Product Type Rayon Rope
Material Natural Fiber
Size 3 mm
For Use With (Application) Reoxygenates faster in bottle, resulting in a drier environment
Quantity 1 Cs.