Learn More
Bellco Glass, Inc. Applikon Mounting Adapter
SDP
Catalog No. 501946207
Encompass_Preferred
This adapter allows the customer to modify an appilkon p-100 motor so that it will fit a bellco 3l or 7l drive stem and vessel. Motor mounting adapter with the following components: Mounting plate a505-632 (1) Mounting pegs a505-617b (4) Flexible motor coupling: A505-612A (1) Bushing A505-624 (1) , 4 screws. MADE IN THE USA
Catalog No. 50-194-6207 Supplier Bellco Glass, Inc. Supplier No. 776480000
This adapter allows the customer to modify an appilkon p-100 motor so that it will fit a bellco 3l or 7l drive stem and vessel. Motor mounting adapter with the following components: Mounting plate a505-632 (1) Mounting pegs a505-617b (4) Flexible motor coupling: A505-612A (1) Bushing A505-624 (1) , 4 screws. MADE IN THE USA