Learn More
Medchemexpress LLC Apraclonidine hydrochloride | 73218-79-8 | 99.0% | 25 MG
SDP
Catalog No. 5000341416
Encompass_Preferred
Apraclonidine hydrochloride is a selective α2 and weak α1 receptor agonist that effectively lowers intraocular pressure (IOP) in human eyes. This topical ophthalmic solution is commonly used for glaucoma, demonstrating lower corneal and blood-brain barrier penetration, which results in fewer adverse systemic effects.
  • Selective α2 and weak α1 receptor agonist
  • Effectively lowers intraocular pressure
  • Functions as a topical ophthalmic solution
  • Can elevate the eye lid
  • Used in glaucoma treatment
  • Exhibits lower corneal and blood-brain barrier penetration
  • Reduces aqueous humor synthesis and has vasoconstrictor actions
Catalog No. 50-003-41416 Supplier Medchemexpress LLC Supplier No. 25MGHY12720A
Apraclonidine hydrochloride is a selective α2 and weak α1 receptor agonist that effectively lowers intraocular pressure (IOP) in human eyes. This topical ophthalmic solution is commonly used for glaucoma, demonstrating lower corneal and blood-brain barrier penetration, which results in fewer adverse systemic effects.
  • Selective α2 and weak α1 receptor agonist
  • Effectively lowers intraocular pressure
  • Functions as a topical ophthalmic solution
  • Can elevate the eye lid
  • Used in glaucoma treatment
  • Exhibits lower corneal and blood-brain barrier penetration
  • Reduces aqueous humor synthesis and has vasoconstrictor actions