Learn More
Medchemexpress LLC HY-P1714A 1mg Medchemexpress, Apraglutide (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1714A1MG
Encompass_Preferred
Medchemexpress, HY-P1714A 1mg Apraglutide (TFA) CAS: Apraglutide TFA (FE 203799 TFA), a synthetic 33-amino-acid peptide and a long-acting GLP-2 analogue, enhances adaptation and linear intestinal growth in a neonatal piglet model of short bowel syndrome with total resection of the ileum [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4241