Learn More
Gold Biotechnology Inc Apramycin Sulfate 10 g
SDP
Catalog No. 5000562173
Encompass_Preferred
Apramycin is an aminoglycoside antibiotic useful against gram-negative bacteria such as E coli and Salmonella species It is used to study antibiotic resistance and protein synthesis translocation inhibition Apramycin sulfate is freely soluble
Catalog No. 50-005-62173 Supplier Gold Biotechnology Inc Supplier No. A60010
Apramycin is an aminoglycoside antibiotic useful against gram-negative bacteria such as E coli and Salmonella species It is used to study antibiotic resistance and protein synthesis translocation inhibition Apramycin sulfate is freely soluble