Learn More
Selleck Chemical LLC Apramycin Sulfate S4254-50mg
SDP
Catalog No. 503111073
Encompass_Preferred
Apramycin Sulfate (Ai3-29795) is an aminoglycoside antibiotic which binds to the deep groove of the RNA
Catalog No. 50-311-1073 Supplier Selleck Chemical LLC Supplier No. S425450MG
Apramycin Sulfate (Ai3-29795) is an aminoglycoside antibiotic which binds to the deep groove of the RNA